trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥97%

Supplier: BeanTown Chemical

Synonyms: 4-(trans-4-Propylcyclohexyl)cyclohexanone

122300-1G 122300-5G
BT122300-1GEA 111 USD
BT122300-1G BT122300-5G
trans-4-(4-n-Propylcyclohexyl)cyclohexanone ≥97%
trans-4-(4-n-Propylcyclohexyl)cyclohexanone

CAS: 82832-73-3; MDL No: MFCD04038797 Liquid; Molecular Formula: C15H26O ; MW: 223.37


  • Liquid

Formula: C₁₅H₂₆O
MW: 222.372 g/mol
MDL Number: MFCD04038797
CAS Number: 82832-73-3

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR