Calcium thioglycolate trihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Calcium mercaptoacetate trihydrate, Calcium thioglycolate trihydrate, Calcium bis(mercaptoacetate) trihydrate, Mercaptoacetic acid calcium salt trihydrate

122275-10G 122275-50G
BT122275-10GEA 29 USD
BT122275-10G BT122275-50G
Calcium thioglycolate trihydrate ≥98%
Calcium bis(mercaptoacetate) (Calcium thioglycolate) trihydrate

CAS: 65208-41-5; EC No: 212-402-5; MDL No: MFCD00150707 Crystalline/Powder; Linear Formula: (SHCH2COO)2Ca·3H2O; MW: 276.34 Melting Point: 270-280°


  • Crystalline/Powder

Formula: C₄H₆CaO₄S₂·3H₂O
MW: 276.35 g/mol
Melting Pt: 270…280 °C
MDL Number: MFCD00150707
CAS Number: 65208-41-5
EINECS: 212-402-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR