2-Fluoro-4-pyridineboronic acid ≥95%

Supplier: BeanTown Chemical

Synonyms: (2-Fluoropyridin-4-yl)boronic acid, 2-Fluoropyridine-4-boronic acid, 2-Fluoro-4-pyridinylboronic acid

122250-1G 122250-5G
BT122250-1GEA 514 USD
BT122250-1G BT122250-5G
2-Fluoro-4-pyridineboronic acid ≥95%
2-Fluoro-4-pyridineboronic acid

CAS: 401815-98-3; MDL No: MFCD04112534 Powder; Molecular Formula: C5H5BFNO2; MW: 140.91


  • Powder

Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
MDL Number: MFCD04112534
CAS Number: 401815-98-3

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR