3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate

Supplier: BeanTown Chemical

122230-250ML 122230-50ML
BT122230-250MLEA 197.47 USD
BT122230-250ML BT122230-50ML
3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate
3,4-Epoxycyclohexylmethyl-3,4-epoxycyclohexanecarboxylate

CAS: 2386-87-0; MDL No: MFCD00440900 Liquid; Molecular Formula: C14H20O4; MW: 252.31 Melting Point: -37°; Boiling Point: 170°; Flash point: 118°C (244°F) Density (g/mL): 1.17; Refractive Index: 1.498


  • Refractive index: 1.498
  • Liquid

Formula: C₁₄H₂₀O₄
Boiling Pt: 170 °C
Melting Pt: –37 °C
Density: 1.17 g/mL
Flash Pt: 118 °C (244 °F)
MDL Number: MFCD00440900
CAS Number: 2386-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR