Rubidium chloride ≥99.95%
Supplier: BeanTown Chemical
Synonyms:
Rubidiumchloride
122200-10G
122200-50G
BT122200-10GEA
289.88
USD
BT122200-10G
BT122200-50G
Rubidium chloride ≥99.95%
Rubidium chloride
CAS: 7791-11-9; EC No: 232-240-9; MDL No: MFCD00011187; RTECS: VL8575000
Crystalline/Powder; Linear Formula: RbCl; MW: 120.92
Melting Point: 715°; Boiling Point: 1390°
Density (g/mL): 2.8; Refractive Index: 1.493
Hygroscopic
- RTECS: VL8575000
- Refractive index: 1.493
- Hygroscopic
- Crystalline/Powder
Formula:
RbCl Boiling Pt: 1390 °C Melting Pt: 715 °C Density: 2.8 g/mL |
MDL Number:
MFCD00011187 CAS Number: 7791-11-9 EINECS: 232-240-9 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...