Erbium(III) oxide ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
Synonyms:
Dierbium trioxide, Erbium oxide
122175-100G
122175-1KG
122175-25G
BT122175-100GEA
196.15
USD
BT122175-100G
BT122175-1KG
BT122175-25G
Erbium(III) oxide ≥99.9% (trace rare earth metals basis)
Erbium(III) oxide
CAS: 12061-16-4; EC No: 235-045-7; MDL No: MFCD00010991
Powder; Molecular Formula: Er2O3; MW: 382.56
Density (g/mL): 8.64
Hygroscopic
- Hygroscopic
- Powder
Formula:
Er₂O₃ MW: 382.52 g/mol Density: 8.64 g/mL |
MDL Number:
MFCD00010991 CAS Number: 12061-16-4 EINECS: 235-045-7 |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...