Triphenylene ≥98%

Supplier: BeanTown Chemical

Synonyms: 9,10-Benzophenanthrene, Isochrysene

122160-1G 122160-5G
BT122160-1GEA 63 USD
BT122160-1G BT122160-5G
Triphenylene ≥98%
Triphenylene

CAS: 217-59-4; EC No: 205-922-9; MDL No: MFCD00001108; RTECS: YK2925000 Solid; Molecular Formula: C18H12; MW: 228.30 Melting Point: 195-198°; Boiling Point: 438°


  • RTECS: YK2925000
  • Solid

Formula: C₁₈H₁₂
MW: 228.29 g/mol
Boiling Pt: 438 °C
Melting Pt: 195…198 °C
MDL Number: MFCD00001108
CAS Number: 217-59-4
EINECS: 205-922-9
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR