3,6-Dichloro-4-methylpyridazine ≥97%
Supplier: BeanTown Chemical
122110-1G
122110-25G
122110-5G
BT122110-1GEA
57
USD
BT122110-1G
BT122110-25G
BT122110-5G
3,6-Dichloro-4-methylpyridazine ≥97%
3,6-Dichloro-4-methylpyridazine
CAS: 19064-64-3; EC No: 242-794-3; MDL No: MFCD00006465
Powder; Molecular Formula: C5H4Cl2N2; MW: 163.00
Melting Point: 86-88°; Boiling Point: 149-151°/21 mmHg
- Powder
Formula:
C₅H₄Cl₂N₂ Boiling Pt: 149…151 °C/21 mmHg Melting Pt: 86…88 °C |
MDL Number:
MFCD00006465 CAS Number: 19064-64-3 EINECS: 242-794-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...