Bromhexine hydrochloride

Supplier: BeanTown Chemical

Synonyms: 2-Amino-3,5-dibromo-N-cyclohexyl-N-methylbenzylamine Hydrochloride, 2,4-Dibromo-6-((cyclohexyl(methyl)amino)methyl)aniline hydrochloride, N-(2-Amino-3,5-dibromobenzyl)-N-methylcyclohexylamine Hydrochloride

122085-100G 122085-500G
BT122085-100GEA 117 USD
BT122085-100G BT122085-500G
Bromhexine hydrochloride
Bromhexine hydrochloride

CAS: 611-75-6; EC No: 210-280-8; MDL No: MFCD00056626; RTECS: XS9950000 Powder; Molecular Formula: C14H20Br2N2·HCl; MW: 412.59


  • RTECS: XS9950000
  • Powder

Formula: C₁₄H₂₀Br₂N₂
MW: 412.59 g/mol
MDL Number: MFCD00056626
CAS Number: 611-75-6
EINECS: 210-280-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR