Melatonin ≥99%
Supplier: BeanTown Chemical
Synonyms:
Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
122070-25G
122070-5G
BT122070-25GEA
163.37
USD
BT122070-25G
BT122070-5G
Melatonin ≥99%
Melatonin
CAS: 73-31-4; EC No: 200-797-7; MDL No: MFCD00005655; RTECS: AC5955000
Powder; Molecular Formula: C13H16N2O2; MW: 232.28
Melting Point: 116.5-118°
- RTECS: AC5955000
- Powder
Formula:
C₁₃H₁₆N₂O₂ MW: 232.28 g/mol Melting Pt: 116.5…118 °C |
MDL Number:
MFCD00005655 CAS Number: 73-31-4 EINECS: 200-797-7 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...