8-Hydroxyjulolidine ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,2,3,5,6,7-Hexahydropyrido[3,2,1-ij]quinolin-8-ol, 2,3,6,7-Tetrahydro-1H,5H-benzo[ij]quinolizin-8-ol

122055-1G 122055-5G
BT122055-1GEA 62 USD
BT122055-1G BT122055-5G
8-Hydroxyjulolidine ≥97%
8-Hydroxyjulolidine

CAS: 41175-50-2; EC No: 255-247-9; MDL No: MFCD00006918 Powder; Molecular Formula: C12H15NO; MW: 189.25 Melting Point: 132-134°


  • Powder

Formula: C₁₂H₁₅NO
MW: 189.26 g/mol
Melting Pt: 132…134 °C
MDL Number: MFCD00006918
CAS Number: 41175-50-2
EINECS: 255-247-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR