L-Glutamic acid 5-benzyl ester ≥99%

Supplier: BeanTown Chemical

Synonyms: γ-Benzyl-L-Glutamate, 5-Benzyl L-Glutamate, gamma-Benzyl L-glutamate, H-Glu(OBzl)-OH

122045-1G 122045-25G 122045-5G
BT122045-1GEA 28 USD
BT122045-1G BT122045-25G BT122045-5G
L-Glutamic acid 5-benzyl ester ≥99%
L-Glutamic acid 5-benzyl ester

CAS: 1676-73-9; EC No: 216-826-1; MDL No: MFCD00002633 Powder; Molecular Formula: C12H15NO4 ; MW: 237.25 Melting Point: 181-182°


  • Powder

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 181…182 °C
MDL Number: MFCD00002633
CAS Number: 1676-73-9
EINECS: 216-826-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR