Triethylene glycol ≥99%

Supplier: BeanTown Chemical

Synonyms: 2,2'-(Ethylenedioxy)diethanol, 2,2'-(Ethane-1,2-diylbis(oxy))diethanol

122020-10L 122020-2.5L 122020-500ML
BT122020-10LEA 250.84 USD
BT122020-10L BT122020-2.5L BT122020-500ML
Triethylene glycol ≥99%
Triethylene glycol

CAS: 112-27-6; EC No: 203-953-2; MDL No: MFCD00002880; RTECS: YE4550000 Liquid; Linear Formula: HO(CH2CH2O)2CH2CH2OH; Molecular Formula: C6H14O4; MW: 150.17 Melting Point: -7°; Boiling Point: 284-288°; Flash point: 166°C (331°F) Density (g/mL): 1.124; Refractive Index: 1.455 Hygroscopic


  • RTECS: YE4550000
  • Refractive index: 1.455
  • Hygroscopic
  • Liquid

Formula: C₆H₁₄O₄
MW: 150.17 g/mol
Boiling Pt: 284…288 °C
Melting Pt: –7 °C
Density: 1.124 g/mL
Flash Pt: 166 °C (331 °F)
MDL Number: MFCD00002880
CAS Number: 112-27-6
EINECS: 203-953-2

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR