Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate

Supplier: BeanTown Chemical

122010-250MG 122010-500MG
BT122010-250MGEA 65 USD
BT122010-250MG BT122010-500MG
Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate
Tris(4,4,4-trifluoro-1-(2-thienyl)-1,3-butanediono)europium(III) hydrate

CAS: 14054-87-6; EC No: 237-892-8; MDL No: MFCD00041948 Powder; Linear Formula: C24H12F9O6S3Eu·xH2O ; Molecular Formula: C24H12EuF9O6S3·xH2O ; MW: 815.51 (anhydrous) Melting Point: 143-144°


  • Powder

Formula: C₂₄H₁₇EuF₉O₇S₃
MW: 851.818 g/mol
Melting Pt: 143…144 °C
MDL Number: MFCD00041948
CAS Number: 14054-87-6
EINECS: 237-892-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR