4-Amino-3-formylpyridine ≥85%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
4-Aminopyridine-3-carboxaldehyde, 4-Aminonicotinaldehyde
122005-1G
122005-5G
BT122005-1GEA
220
USD
BT122005-1G
BT122005-5G
4-Amino-3-formylpyridine ≥85%, Technical Grade
4-Amino-3-formylpyridine
CAS: 42373-30-8; MDL No: MFCD04038972
Solid; Molecular Formula: C6H6N2O; MW: 122.12
Melting Point: 113-118°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₆H₆N₂O MW: 122.13 Melting Pt: 113…118 °C |
MDL Number:
MFCD04038972 CAS Number: 42373-30-8 |
Specification Test Results
Assay | 85% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...