3-Quinolinecarboxylic acid ≥98%

Supplier: BeanTown Chemical

122000-1G 122000-5G
BT122000-1GEA 55 USD
BT122000-1G BT122000-5G
3-Quinolinecarboxylic acid ≥98%
3-Quinolinecarboxylic acid

CAS: 6480-68-8; EC No: 229-337-3; MDL No: MFCD00006770 Powder; Molecular Formula: C10H6NO2; MW: 173.17 Melting Point: 277-280°


  • Powder

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Melting Pt: 277…280 °C
MDL Number: MFCD00006770
CAS Number: 6480-68-8
EINECS: 229-337-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR