Iron(II) phthalocyanine ≥96%

Supplier: BeanTown Chemical

Synonyms: Phthalocyanine Iron(II), Iron phthalocyanine

121995-1G 121995-5G
BT121995-1GEA 29 USD
BT121995-1G BT121995-5G
Iron(II) phthalocyanine ≥96%
Iron(II) phthalocyanine

CAS: 132-16-1; EC No: 205-047-2; MDL No: MFCD00015953 Crystalline/Powder; Molecular Formula: C32H16FeN8 ; MW: 568.3


  • Crystalline/Powder

Formula: C₃₂H₁₆FeN₈
MW: 568.38 g/mol
MDL Number: MFCD00015953
CAS Number: 132-16-1
EINECS: 205-047-2

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR