Calcium-D-gluconate monohydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate

121990-100G 121990-2.5KG 121990-500G
BT121990-100GEA 24 USD
BT121990-100G BT121990-2.5KG BT121990-500G
Calcium-D-gluconate monohydrate ≥98%
Calcium-D-gluconate monohydrate

CAS: 299-28-5; EC No: 206-075-8; MDL No: MFCD00064209; RTECS: EW2100000 Powder; Molecular Formula: C12H22CaO14·H2O ; MW: 448.39

Formula: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Melting Pt: 178
MDL Number: MFCD00150787
CAS Number: 66905-23-5
EINECS: 206-075-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR