3'-Fluoropropiophenone ≥97%
Supplier: BeanTown Chemical
121930-1G
121930-5G
BT121930-1GEA
62
USD
BT121930-1G
BT121930-5G
3'-Fluoropropiophenone ≥97%
3'-Fluoropropiophenone
CAS: 455-67-4; MDL No: MFCD00061245
Liquid; Molecular Formula: C9H9FO; MW: 152.17
Boiling Point: 94-96°/5 mmHg
Refractive Index: 1.505
- Refractive index: 1.505
- Liquid
Formula:
C₉H₉FO Boiling Pt: 94…96 °C/5 mmHg |
MDL Number:
MFCD00061245 CAS Number: 455-67-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...