1-Heptanesulphonic acid sodium salt monohydrate ≥99%

Supplier: BeanTown Chemical

Synonyms: Sodium 1-heptanesulphonate monohydrate

121865-10G 121865-50G
BT121865-10GEA 53 USD
BT121865-10G BT121865-50G
1-Heptanesulphonic acid sodium salt monohydrate ≥99%
1-Heptanesulphonic acid sodium salt monohydrate

CAS: 207300-90-1; EC No: 245-210-5; MDL No: MFCD00149550 Solid; Linear Formula: CH3(CH2)6SO3Na·H2O; Molecular Formula: C7H15NaO3S·H2O; MW: 220.26 Melting Point: 300°


  • Solid

Formula: C₇H₁₅NaO₃S·1H₂O
Melting Pt: >300 °C
MDL Number: MFCD00149550
CAS Number: 207300-90-1
EINECS: 245-210-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR