Ytterbium (III) trifluoromethanesulfonate hydrate (∼25 - 28% Yb)

Supplier: BeanTown Chemical
Warning

Synonyms: Trifluoromethanesulphonic acid ytterbium salt hydrate, Ytterbium(III) trifluoromethanesulphonate hydrate, Ytterbium(III) triflate hydrate, Ytterbium triflate hydrate

121855-25G 121855-5G
BT121855-25GEA 299 USD
BT121855-25G BT121855-5G
Ytterbium (III) trifluoromethanesulfonate hydrate (∼25 - 28% Yb)
Ytterbium(III) trifluoromethanesulphonate hydrate

MDL No: MFCD03703499 Melting Point: 118-122° Hygroscopic


  • Hygroscopic

Formula: C₃F₉O₉S₃Yb·xH₂O
MW: 620.25 g/mol (anhy)
Melting Pt: 118…122 °C
MDL Number: MFCD03703499
CAS Number: 252976-51-5

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR