Formamidine acetate ≥99%

Supplier: BeanTown Chemical

121850-100G 121850-500G
BT121850-100GEA 49 USD
BT121850-100G BT121850-500G
Formamidine acetate ≥99%
Formamidine acetate

CAS: 3473-63-0; EC No: 222-442-5; MDL No: MFCD00012866 Crystalline; Linear Formula: HN=CHNH2·CH3COOH; Molecular Formula: C3H8N2O2; MW: 104.11 Melting Point: 158-161° (decomposes) Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: C₃H₈N₂O₂
Melting Pt: 158…161 °C (dec.)
MDL Number: MFCD00012866
CAS Number: 3473-63-0
EINECS: 222-442-5

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR