4-((tert-Butoxycarbonyl)amino)benzoic acid ≥97%

Supplier: BeanTown Chemical

Synonyms: Boc-4-Abz-OH, 4-(Boc-amino)benzoic Acid

121815-1G 121815-5G
BT121815-1GEA 37.55 USD
BT121815-1G BT121815-5G
4-((tert-Butoxycarbonyl)amino)benzoic acid ≥97%
4-((tert-Butoxycarbonyl)amino)benzoic acid

CAS: 66493-39-8; MDL No: MFCD00037428 Powder; Molecular Formula: C12H15NO4; MW: 237.26 Melting Point: 200° (decomposes)


  • Powder

Formula: C₁₂H₁₅NO₄
MW: 237.3 g/mol
Melting Pt: 200 °C (dec.)
MDL Number: MFCD00037428
CAS Number: 66493-39-8

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR