N-n-Octyl-D-glucamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
(2R,3R,4R,5S)-6-(Octylamino)hexane-1,2,3,4,5-pentaol, 1-Deoxy-1-(n-octylamino)-D-glucitol
121810-1G
121810-250MG
BT121810-1GEA
59
USD
BT121810-1G
BT121810-250MG
N-n-Octyl-D-glucamine ≥98%
N-n-Octyl-D-glucamine
CAS: 23323-37-7; EC No: 245-582-9; MDL No: MFCD00134352
Solid; Molecular Formula: C14H31NO5; MW: 293.40
Melting Point: 121-124°
Optical Rotation: [α]20/D -15°, c = 1 in methanol
- Optical rotation: [α]20/D -15°, c = 1 in methanol
- Solid
Formula:
C₁₄H₃₁NO₅ MW: 293.40 g/mol Melting Pt: 121…124 °C |
MDL Number:
MFCD00134352 CAS Number: 23323-37-7 EINECS: 245-582-9 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...