α-Tropolone ≥98%

Supplier: BeanTown Chemical

121805-1G 121805-25G 121805-5G
BT121805-1GEA 38 USD
BT121805-1G BT121805-25G BT121805-5G
α-Tropolone ≥98%
α-Tropolone

CAS: 533-75-5; EC No: 208-577-2; MDL No: MFCD00004158; RTECS: GU4075000 Crystalline; Molecular Formula: C7H6O2; MW: 122.12 Melting Point: 50-52°; Boiling Point: 80-84°/0.1 mmHg; Flash point: 112°C (234°F) Hygroscopic


  • RTECS: GU4075000
  • Hygroscopic
  • Crystalline

Formula: C₇H₆O₂
Boiling Pt: 80…84 °C/0.1 mmHg
Melting Pt: 50…52 °C
Flash Pt: 112 °C (234 °F)
MDL Number: MFCD00004158
CAS Number: 533-75-5
EINECS: 208-577-2

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR