Bis(2-ethylhexyl)adipate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
121800-100G
121800-25G
BT121800-100GEA
120.04
USD
BT121800-100G
BT121800-25G
Bis(2-ethylhexyl)adipate ≥99%
Bis(2-ethylhexyl)adipate
CAS: 103-23-1; EC No: 203-090-1; MDL No: MFCD00009496; RTECS: AU9700000
Liquid; Molecular Formula: C22H42O4; MW: 370.57
Melting Point: -70°; Boiling Point: 175°/2 mmHg; Flash point: 196°C (385°F)
Density (g/mL): 0.925; Refractive Index: 1.447
- RTECS: AU9700000
- Refractive index: 1.447
- Liquid
Formula:
C₂₂H₄₂O₄ Boiling Pt: 175 °C/2 mmHg Melting Pt: –70 °C Density: 0.925 g/mL Flash Pt: 196 °C (385 °F) |
MDL Number:
MFCD00009496 CAS Number: 103-23-1 EINECS: 203-090-1 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...