Methyl (S)-(+)-2-pyrrolidone-5-carboxylate ≥95%
Supplier: BeanTown Chemical
Synonyms:
Methyl (S)-(+)-2-oxopyrrolidine-5-carboxylate, H-Pyr-OMe
121780-100G
121780-25G
121780-5G
BT121780-100GEA
940
USD
BT121780-100G
BT121780-25G
BT121780-5G
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate ≥95%
Methyl (S)-(+)-2-pyrrolidone-5-carboxylate
CAS: 4931-66-2; EC No: 225-567-3; MDL No: MFCD00080931
Liquid; Molecular Formula: C6H9NO3; MW: 143.14
Boiling Point: 90°/0.3 mmHg; Flash point:
- Optical rotation: [α]20/D +10.5°, c = 1 in ethanol
- Refractive index: 1.486
- Liquid
Formula:
C₆H₉NO₃ MW: 143.14 g/mol Boiling Pt: 90 °C/0.3 mmHg Density: 1.226 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00080931 CAS Number: 4931-66-2 EINECS: 225-567-3 |
Specification Test Results
Assay | 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...