Dibenzothiophene-4-boronic acid ≥95%
Supplier: BeanTown Chemical
121760-1G
121760-5G
BT121760-1GEA
56
USD
BT121760-1G
BT121760-5G
Dibenzothiophene-4-boronic acid ≥95%
Dibenzothiophene-4-boronic acid
CAS: 108847-20-7; MDL No: MFCD01318182
Powder; Molecular Formula: C12H9BO2S; MW: 228.08
Melting Point: 327-330°
- Powder
Formula:
C₁₂H₉BO₂S Melting Pt: 327…330 °C |
MDL Number:
MFCD01318182 CAS Number: 108847-20-7 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...