Phenolphthalein ACS

Supplier: BeanTown Chemical

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

121735-100G 121735-500G
BT121735-100GEA 33.66 USD
BT121735-100G BT121735-500G
Phenolphthalein ACS
Phenolphthalein

CAS: 77-09-8; EC No: 201-004-7; MDL No: MFCD00005913; RTECS: SM8380000 Powder; Molecular Formula: C20H14O4 ; MW: 318.33 Melting Point: 261-263° Density (g/mL): 1.299


  • RTECS: SM8380000
  • Powder

Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Melting Pt: 261…263 °C
Density: 1.299 g/mL
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Clarity of solution Passes test
Visual transition pH 8.0-10
Color Colorless to red

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR