2,3-Dichlorobenzoic acid ≥98%
Supplier: BeanTown Chemical
121715-10G
121715-50G
BT121715-10GEA
71
USD
BT121715-10G
BT121715-50G
2,3-Dichlorobenzoic acid ≥98%
2,3-Dichlorobenzoic acid
CAS: 50-45-3; EC No: 200-039-5; MDL No: MFCD00002413; RTECS: DG6475000
Powder; Molecular Formula: C7H3Cl2O2; MW: 191.01
Melting Point: 168-170°
- RTECS: DG6475000
- Powder
Formula:
C₇H₄Cl₂O₂ MW: 191.01 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD00002413 CAS Number: 50-45-3 EINECS: 200-039-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...