4-Chlorobenzoylacetonitrile ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Chlorophenacylcyanide, 3-(4-Chlorophenyl)-3-oxopropanenitrile

121710-100G 121710-25G 121710-5G
BT121710-100GEA 825 USD
BT121710-100G BT121710-25G BT121710-5G
4-Chlorobenzoylacetonitrile ≥98%
4-Chlorobenzoylacetonitrile

CAS: 4640-66-8; MDL No: MFCD00051625 Solid; Molecular Formula: C9H6ClNO; MW: 179.60 Melting Point: 129-133°


  • Solid

Formula: C₉H₆ClNO
MW: 179.61 g/mol
Melting Pt: 129…133 °C
MDL Number: MFCD00051625
CAS Number: 4640-66-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR