Tetramethylammonium formate 30% (w/w) in water

Supplier: BeanTown Chemical

121690-100ML 121690-500ML
BT121690-100MLEA 78.94 USD
BT121690-100ML BT121690-500ML
Tetramethylammonium formate 30% (w/w) in water
Tetramethylammonium formate

CAS: 59138-84-0; EC No: 261-622-8; MDL No: MFCD00054402; RTECS: BS7960000 Liquid; Linear Formula: (CH3)4N(HCOO); Molecular Formula: C5H13NO2; MW: 119.16 Boiling Point: 102° Density (g/mL): 1.008; Refractive Index: 1.364


  • RTECS: BS7960000
  • Refractive index: 1.364
  • Liquid

Formula: C₅H₁₃NO₂
Boiling Pt: 102 °C
Density: 1.008 g/mL
MDL Number: MFCD00054402
CAS Number: 59138-84-0
EINECS: 261-622-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR