3-Pyridone ≥98%

Supplier: BeanTown Chemical

Synonyms: 3-Hydroxypyridine, β-Pyridone, Pyridin-3-ol, 3-Pyridinol

121640-100G 121640-500G 121640-50G
BT121640-100GEA 49 USD
BT121640-100G BT121640-500G BT121640-50G
3-Pyridone ≥98%
3-Pyridone

CAS: 109-00-2; EC No: 205-520-3; MDL No: MFCD00006378; RTECS: UV1144050 Crystalline; Molecular Formula: C5H5NO; MW: 95.10 Melting Point: 125-128°


  • RTECS: UV1144050
  • Crystalline

Formula: C₅H₅NO
MW: 95.1 g/mol
Melting Pt: 125…128 °C
MDL Number: MFCD00006378
CAS Number: 109-00-2
EINECS: 205-520-3

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR