Tris(triphenylphosphine)ruthenium(II) chloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
Dichlorotris(triphenylphosphine)ruthenium(II), Dichlorotris(triphenylphosphino)ruthenium (II)
121635-1G
121635-25G
121635-5G
BT121635-1GEA
37
USD
BT121635-1G
BT121635-25G
BT121635-5G
Tris(triphenylphosphine)ruthenium(II) chloride ≥97%
Tris(triphenylphosphine)ruthenium(II) chloride
CAS: 15529-49-4; EC No: 239-569-7; MDL No: MFCD00013077
Crystalline; Linear Formula: [(C6H5)3P]3RuCl2; Molecular Formula: C54H45Cl2P3Ru; MW: 958.83
Melting Point: 159°
Air Sensitive, Moisture Sensitive
- Air Sensitive, Moisture Sensitive
- Crystalline
Formula:
C₅₄H₄₅Cl₂P₃Ru Melting Pt: 159 °C |
MDL Number:
MFCD00013077 CAS Number: 15529-49-4 EINECS: 239-569-7 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...