(2,4-Dichlorobenzyl)triphenylphosphonium chloride
Supplier: BeanTown Chemical
Synonyms:
Triphenyl(2,4-dichlorobenzyl)phosphonium Chloride
121630-100G
121630-25G
BT121630-100GEA
365
USD
BT121630-100G
BT121630-25G
(2,4-Dichlorobenzyl)triphenylphosphonium chloride
(2,4-Dichlorobenzyl)triphenylphosphonium chloride
CAS: 2492-23-1; EC No: 219-659-2; MDL No: MFCD00031641; RTECS: TA2200000
Solid; Molecular Formula: C25H20Cl3P; MW: 457.76
- RTECS: TA2200000
- Solid
Formula:
C₂₅H₂₀Cl₃P MW: 457.76 g/mol |
MDL Number:
MFCD00031641 CAS Number: 2492-23-1 EINECS: 219-659-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...