Lithium carbonate ≥99.0% ACS
Supplier: BeanTown Chemical
121610-2KG
121610-500G
BT121610-2KGEA
642
USD
BT121610-2KG
BT121610-500G
Lithium carbonate ≥99.0% ACS
Lithium carbonate
CAS: 554-13-2; EC No: 209-062-5; MDL No: MFCD00011084
Powder; Linear Formula: Li2CO3; MW: 73.90
Melting Point: 618°
- Powder
Formula:
Li₂CO₃ Melting Pt: 618 °C |
MDL Number:
MFCD00011084 CAS Number: 554-13-2 EINECS: 209-062-5 |
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...