D-(-)-Serine ≥99%

Supplier: BeanTown Chemical

Synonyms: (R)-(-)-Serine, H-D-Ser-OH

121585-100G 121585-25G 121585-5G
BT121585-100GEA 385 USD
BT121585-100G BT121585-25G BT121585-5G
D-(-)-Serine ≥99%
D-(-)-Serine

CAS: 312-84-5; EC No: 206-229-4; MDL No: MFCD00004269; RTECS: VT8200000 Crystalline; Molecular Formula: C3H7NO3; MW: 105.09


  • RTECS: VT8200000
  • Crystalline

Formula: C₃H₇NO₃
MW: 105.09 g/mol
Melting Pt: 235
MDL Number: MFCD00004269
CAS Number: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR