2-(Diphenylphosphino)benzaldehyde ≥97%

Supplier: BeanTown Chemical

Synonyms: (2-Formylphenyl)diphenylphosphine

121565-1G 121565-5G
BT121565-1GEA 71 USD
BT121565-1G BT121565-5G
2-(Diphenylphosphino)benzaldehyde ≥97%
2-(Diphenylphosphino)benzaldehyde

CAS: 50777-76-9; MDL No: MFCD00013367 Solid; Molecular Formula: C19H15OP; MW: 290.30 Melting Point: 112-115°


  • Solid

Formula: C₁₉H₁₅OP
MW: 290.30 g/mol
Melting Pt: 112…115 °C
MDL Number: MFCD00013367
CAS Number: 50777-76-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR