L-Prolinamide ≥99%
Supplier: BeanTown Chemical
Synonyms:
H-Pro-NH₂, (S)-2-Pyrrolidinecarboxamide, H-Pro-NH2
121545-1G
121545-5G
BT121545-1GEA
73
USD
BT121545-1G
BT121545-5G
L-Prolinamide ≥99%
L-Prolinamide
CAS: 7531-52-4; EC No: 231-397-0; MDL No: MFCD00005253
Powder; Molecular Formula: C5H10N2O ; MW: 114.15
Melting Point: 95-97°
- Powder
Formula:
C₅H₁₀N₂O MW: 114.15 g/mol Melting Pt: 95…97 °C |
MDL Number:
MFCD00005253 CAS Number: 7531-52-4 EINECS: 231-397-0 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...