Silver ≥99.99% (trace metals basis), slug, Length: 6.35 mm, Diameter 6.35 mm

Supplier: BeanTown Chemical

121520-25SLUG 121520-5SLUG
BT121520-25SLUEA 209 USD
BT121520-25SLU BT121520-5SLUG
Silver ≥99.99% (trace metals basis), slug, Length: 6.35 mm, Diameter 6.35 mm
Silver

CAS: 7440-22-4; EC No: 231-131-3; MDL No: MFCD00003397 Solid; Molecular Formula: Ag; MW: 107.87 Melting Point: 960°; Boiling Point: 2212° Density (g/mL): 10.49


  • Solid

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.49 g/mL
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR