Indium tin oxide (In₂O₃:SnO₂) (90:10%) (w/w) ≥99.99% (trace metals basis), pieces 3-12mm
Supplier: BeanTown Chemical
Synonyms:
ITO
121500-100G
121500-25G
121500-5G
BT121500-100GEA
695
USD
BT121500-100G
BT121500-25G
BT121500-5G
Indium tin oxide (In₂O₃:SnO₂) (90:10%) (w/w) ≥99.99% (trace metals basis), pieces 3-12mm
Indium tin oxide
CAS: 50926-11-9; MDL No: MFCD00171662
3-12mm Pieces
Melting Point: 1910°
Formula:
In₂O₃·(SnO₂)x Melting Pt: 1910 °C |
MDL Number:
MFCD00171662 CAS Number: 50926-11-9 |
Specification Test Results
Assay | 99.99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...