Ethylammonium chloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Aminoethane hydrochloride, Ethylamine hydrochloride
121475-1KG
121475-250G
BT121475-1KGEA
303
USD
BT121475-1KG
BT121475-250G
Ethylammonium chloride ≥98%
Ethylammonium chloride
CAS: 557-66-4; EC No: 209-182-8; MDL No: MFCD00012885; RTECS: KH2495000
Crystalline; Linear Formula: CH3CH2NH2·HCl; Molecular Formula: C2H7N·HCl; MW: 81.55
Melting Point: 107-108°
Hygroscopic
- RTECS: KH2495000
- Hygroscopic
- Crystalline
Formula:
C₂H₈ClN Melting Pt: 107…108 °C |
MDL Number:
MFCD00012885 CAS Number: 557-66-4 EINECS: 209-182-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...