Samarium(III) oxide ≥99.9%
Supplier: BeanTown Chemical
121435-100G
121435-25G
121435-500G
BT121435-100GEA
173
USD
BT121435-100G
BT121435-25G
BT121435-500G
Samarium(III) oxide ≥99.9%
Samarium(III) oxide
CAS: 12060-58-1; EC No: 235-043-6; MDL No: MFCD00011237
Powder; Linear Formula: Sm2O3; MW: 348.72
Melting Point: 2325°
Density (g/mL): 8.35; Refractive Index: 1.97
- Refractive index: 1.97
- Powder
Formula:
O₃Sm₂ Melting Pt: 2325 °C Density: 8.35 g/mL |
MDL Number:
MFCD00011237 CAS Number: 12060-58-1 EINECS: 235-043-6 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...