Lutetium trichloride, anhydrous ≥99.9%

Supplier: BeanTown Chemical

121425-1G 121425-5G
BT121425-1GEA 118 USD
BT121425-1G BT121425-5G
Lutetium trichloride, anhydrous ≥99.9%
Lutetium trichloride

CAS: 10099-66-8; EC No: 233-240-1; MDL No: MFCD00011099 -20 Mesh Powder; Linear Formula: LuCl3; MW: 281.33 Melting Point: 905° Density (g/mL): 3.98 Hygroscopic


  • Hygroscopic
  • -20 Mesh Powder

Formula: Cl₃Lu
MW: 281.33
Melting Pt: 892
Density: 3.98 g/mL
MDL Number: MFCD00011099
CAS Number: 10099-66-8
EINECS: 233-240-1

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR