5-Quinolinol ≥99%
Supplier: BeanTown Chemical
Synonyms:
5-Hydroxyquinoline, Quinolin-5-ol
121415-1G
121415-250MG
BT121415-1GEA
176
USD
BT121415-1G
BT121415-250MG
5-Quinolinol ≥99%
5-Quinolinol
CAS: 578-67-6; EC No: 209-428-4; MDL No: MFCD00006792; RTECS: VC4100000
Crystalline; Molecular Formula: C9H7NO ; MW: 145.16
Melting Point: 223-226°
- RTECS: VC4100000
- Crystalline
Formula:
C₉H₇NO MW: 145.16 g/mol Melting Pt: 223…226 °C |
MDL Number:
MFCD00006792 CAS Number: 578-67-6 EINECS: 209-428-4 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...