Niobium(V) oxide ≥99.9%
Supplier: BeanTown Chemical
Synonyms:
Diniobium pentaoxide, Niobium pentoxide
121410-100G
121410-500G
BT121410-100GEA
100.66
USD
BT121410-100G
BT121410-500G
Niobium(V) oxide ≥99.9%
Niobium(V) oxide
CAS: 1313-96-8; EC No: 215-213-6; MDL No: MFCD00011128
Powder; Molecular Formula: Nb2O5; MW: 265.81
Melting Point: 1520°
Density (g/mL): 4.47
- Powder
Formula:
Nb₂O₅ Melting Pt: 1520 °C Density: 4.47 g/mL |
MDL Number:
MFCD00011128 CAS Number: 1313-96-8 EINECS: 215-213-6 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...