cis-1,4-Diacetoxy-2-butene ≥95%
Supplier: BeanTown Chemical
CAS: 25260-60-0; MDL No: MFCD00059339
Liquid; Linear Formula: CH3CO2CH2CH=CHCH2O2CCH3; Molecular Formula: C8H12O4; MW: 172.18
Boiling Point: 120-121°/18 mmHg; Flash point:
- Refractive index: 1.443
- Liquid
Formula:
C₈H₁₂O₄ Boiling Pt: 120…121 °C/18 mmHg Density: 1.08 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00059339 CAS Number: 25260-60-0 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...