tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%

Supplier: BeanTown Chemical

Synonyms: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide

121395-1G 121395-25G 121395-5G
BT121395-1GEA 29 USD
BT121395-1G BT121395-25G BT121395-5G
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate

CAS: 152120-54-2; MDL No: MFCD01075122 Solid; Molecular Formula: C14H22N4O4 ; MW: 310.35 Melting Point: 86-90°


  • Solid

Formula: C₁₄H₂₂N₄O₄
MW: 310.354 g/mol
Melting Pt: 86…90 °C
MDL Number: MFCD01075122
CAS Number: 152120-54-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR