2-Amino-1-naphthalenesulfonic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Aminonaphthalene-1-sulphonic acid
121380-2.5KG
121380-500G
BT121380-2.5KGEA
248
USD
BT121380-2.5KG
BT121380-500G
2-Amino-1-naphthalenesulfonic acid ≥98%
2-Amino-1-naphthalenesulphonic acid
CAS: 81-16-3; EC No: 201-331-5; MDL No: MFCD00003988; RTECS: QK1250000
Crystalline; Molecular Formula: C10H9NO3S; MW: 223.25
- RTECS: QK1250000
- Crystalline
Formula:
C₁₀H₉NO₃S MW: 223.25 g/mol |
MDL Number:
MFCD00003988 CAS Number: 81-16-3 EINECS: 201-331-5 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...