4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥97%

Supplier: BeanTown Chemical

Synonyms: trans,trans-4'-Pentylbicyclohexyl-4-carboxylic acid 4-Fluorophenyl ester, 4-Fluorophenyl trans,trans-4'-Pentylbicyclohexyl-4-carboxylate, (Trans,trans)-4-fluorophenyl 4'-pentyl-[1,1'-bi(cyclohexane)]-4-carboxylate

121360-1G 121360-5G
BT121360-1GEA 103 USD
BT121360-1G BT121360-5G
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate ≥97%
4-Fluorophenyl-trans,trans-4'-n-pentylbicyclohexyl-4-carboxylate

CAS: 88878-50-6; MDL No: MFCD11053423 Solid; Molecular Formula: C24H35FO2; MW: 374.54 Melting Point: 64-66°


  • Solid

Formula: C₂₄H₃₅FO₂
MW: 374.54 g/mol
Melting Pt: 64…66 °C
MDL Number: MFCD11053423
CAS Number: 88878-50-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR